Assay Details
| Gene Symbol: ARL2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692017.3 | XP_005692074.1 | 4 - 5 | 468 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARL2BP| Gene Symbol: ARL2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692017.3 | XP_005692074.1 | 4 - 5 | 468 | 65 | |
Set Membership: |
|