Assay Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697552.3 | XP_005697609.2 | 4 - 5 | 657 | 58 | |
| XM_005697553.3 | XP_005697610.2 | 4 - 5 | 619 | 58 | ||
| XM_005697554.3 | XP_005697611.2 | 3 - 4 | 505 | 58 | ||
| XM_018041024.1 | XP_017896513.1 | 3 - 4 | 505 | 58 | ||
| GenBank mRNA | JO419569.2 | - | 4 - 5 | 870 | 58 | |
| JO438726.2 | - | 3 - 4 | 508 | 58 | ||
More Information
Set Membership: |
|

ACSM2B
Genomic Map