Assay Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697552.3 | XP_005697609.2 | 8 - 9 | 1248 | 80 | |
| XM_005697553.3 | XP_005697610.2 | 8 - 9 | 1210 | 80 | ||
| XM_005697554.3 | XP_005697611.2 | 7 - 8 | 1096 | 80 | ||
| XM_018041024.1 | XP_017896513.1 | 7 - 8 | 1096 | 80 | ||
| GenBank mRNA | JO438726.2 | - | 7 - 8 | 1099 | 80 | |
More Information
Set Membership: |
|

ACSM2B
Genomic Map