Assay Details
| Gene Symbol: TCTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698262.3 | XP_005698319.2 | 2 - 3 | 359 | 66 | |
| XM_013975159.2 | XP_013830613.2 | 2 - 3 | 462 | 66 | ||
| GenBank mRNA | JT691760.1 | - | 16 - 17 | 2026 | 66 | |
More Information
Set Membership: |
|

TCTN3
Genomic Map