Assay Details
| Gene Symbol: LOC102168979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043004.1 | XP_017898493.1 | 2 - 3 | 422 | 69 | |
| GenBank mRNA | JO422175.2 | - | 2 - 3 | 541 | 69 | |
More Information
Set Membership: |
|

LOC102168979
Genomic Map