Assay Details
| Gene Symbol: HTATSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044215.1 | XP_017899704.1 | 9 - 10 | 1310 | 65 | |
| GenBank mRNA | JO496552.2 | - | 1011 | 65 | ||
More Information
Set Membership: |
|

HTATSF1
Genomic Map