Assay Details
| Gene Symbol: LOC102176107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700437.2 | XP_005700494.1 | 2 - 3 | 287 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102176107| Gene Symbol: LOC102176107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700437.2 | XP_005700494.1 | 2 - 3 | 287 | 68 | |
Set Membership: |
|