Assay Details
| Gene Symbol: PDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043835.1 | XP_017899324.1 | 5 - 6 | 1119 | 65 | |
| XM_018043836.1 | XP_017899325.1 | 5 - 6 | 1119 | 65 | ||
| GenBank mRNA | KA330984.1 | - | 7 - 8 | 1324 | 65 | |
More Information
Set Membership: |
|

PDK3
Genomic Map