Assay Details
| Gene Symbol: LOC102177044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043707.1 | XP_017899196.1 | 4 - 5 | 335 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102177044| Gene Symbol: LOC102177044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043707.1 | XP_017899196.1 | 4 - 5 | 335 | 104 | |
Set Membership: |
|