Assay Details
Gene Symbol: LOC102173650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049918.1 | XP_017905407.1 | 4 - 5 | 1308 | 74 | |
GenBank mRNA | JT653721.1 | - | 9 - 10 | 2921 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
