Assay Details
| Gene Symbol: PGLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051386.1 | XP_017906875.1 | 4 - 5 | 843 | 58 | |
| GenBank mRNA | JO613616.2 | - | 4 - 5 | 716 | 58 | |
| KA324712.1 | - | 1 - 2 | 977 | 58 | ||
More Information
Set Membership: |
|

PGLS
Genomic Map