Assay Details
| Gene Symbol: SLC6A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050160.1 | XP_017905649.1 | 5 - 6 | 990 | 62 | |
| XM_018050161.1 | XP_017905650.1 | 5 - 6 | 990 | 62 | ||
| GenBank mRNA | JO637335.2 | - | 5 - 6 | 1013 | 62 | |
More Information
Set Membership: |
|

SLC6A7
Genomic Map