Assay Details
| Gene Symbol: MXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055462.1 | XP_017910951.1 | 5 - 6 | 729 | 72 | |
| XM_018055463.1 | XP_017910952.1 | 5 - 6 | 1296 | 72 | ||
| GenBank mRNA | KA311231.1 | - | 5 - 6 | 759 | 72 | |
More Information
Set Membership: |
|

MXD1
Genomic Map