Assay Details
| Gene Symbol: WBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056809.1 | XP_017912298.1 | 4 - 5 | 600 | 66 | |
| XM_018056810.1 | XP_017912299.1 | 4 - 5 | 354 | 66 | ||
| XM_018056812.1 | XP_017912301.1 | 3 - 4 | 1073 | 66 | ||
More Information
Set Membership: |
|

WBP4
Genomic Map