Assay Details
| Gene Symbol: CRYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056536.1 | XP_017912025.1 | 7 - 8 | 936 | 65 | |
| GenBank mRNA | JO434945.2 | - | 1 - 2 | 557 | 65 | |
| JO576900.1 | - | 7 - 8 | 955 | 65 | ||
| KA319977.1 | - | 1 - 2 | 606 | 65 | ||
More Information
Set Membership: |
|

CRYL1
Genomic Map