Assay Details
| Gene Symbol: MEAK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061792.1 | XP_017917281.1 | 3 - 4 | 682 | 55 | |
| XM_018061793.1 | XP_017917282.1 | 4 - 5 | 658 | 55 | ||
| XM_018061794.1 | XP_017917283.1 | 4 - 5 | 688 | 55 | ||
More Information
Set Membership: |
|

MEAK7
Genomic Map