Assay Details
| Gene Symbol: FOSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062603.1 | XP_017918092.1 | 2 - 3 | 998 | 72 | |
| XM_018062604.1 | XP_017918093.1 | 2 - 3 | 1000 | 72 | ||
| GenBank mRNA | FJ455501.1 | - | 2 - 3 | 456 | 72 | |
| FJ493226.1 | - | 1 - 2 | 235 | 72 | ||
More Information
Set Membership: |
|

FOSB
Genomic Map