Assay Details
| Gene Symbol: LOC108633778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039360.1 | XP_017894849.1 | 4 - 5 | 594 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC108633778| Gene Symbol: LOC108633778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039360.1 | XP_017894849.1 | 4 - 5 | 594 | 63 | |
Set Membership: |
|