Assay Details
| Gene Symbol: LOC102180560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039865.1 | XP_017895354.1 | 25 - 26 | 4069 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102180560| Gene Symbol: LOC102180560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039865.1 | XP_017895354.1 | 25 - 26 | 4069 | 96 | |
Set Membership: |
|