Assay Details
| Gene Symbol: LOC102181259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974863.2 | XP_013830317.2 | 2 - 3 | 830 | 69 | |
| GenBank mRNA | JO422551.2 | - | 2 - 3 | 520 | 69 | |
| JO605549.2 | - | 9 - 10 | 1476 | 69 | ||
More Information
Set Membership: |
|

LOC102181259
Genomic Map