Assay Details
| Gene Symbol: LOC102181259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO422551.2 | - | 4 - 5 | 757 | 78 | |
| JO605549.2 | - | 7 - 8 | 1239 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102181259| Gene Symbol: LOC102181259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO422551.2 | - | 4 - 5 | 757 | 78 | |
| JO605549.2 | - | 7 - 8 | 1239 | 78 | ||
Set Membership: |
|