Assay Details
| Gene Symbol: LOC102181259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974863.2 | XP_013830317.2 | 6 - 7 | 1330 | 93 | |
| GenBank mRNA | JO422551.2 | - | 6 - 7 | 1020 | 93 | |
| JO605549.2 | - | 5 - 6 | 976 | 93 | ||
More Information
Set Membership: |
|

LOC102181259
Genomic Map