Assay Details
| Gene Symbol: LOC102181259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974863.2 | XP_013830317.2 | 8 - 9 | 1535 | 79 | |
| GenBank mRNA | JO422551.2 | - | 8 - 9 | 1225 | 79 | |
| JO605549.2 | - | 3 - 4 | 771 | 79 | ||
More Information
Set Membership: |
|

LOC102181259
Genomic Map