Assay Details
| Gene Symbol: KRTCAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046232.1 | XP_017901721.1 | 4 - 5 | 383 | 68 | |
| GenBank mRNA | JO445826.2 | - | 222 | 68 | ||
| JO581641.1 | - | 4 - 5 | 521 | 68 | ||
More Information
Set Membership: |
|

KRTCAP2
Genomic Map