Assay Details
| Gene Symbol: HSPA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674769.3 | XP_005674826.1 | 4 - 5 | 798 | 97 | |
| GenBank mRNA | JO358407.2 | - | 1 - 2 | 603 | 97 | |
| JO406248.2 | - | 1 - 2 | 761 | 97 | ||
| JO430237.2 | - | 1 - 2 | 1283 | 97 | ||
| JO587094.2 | - | 1 - 2 | 1287 | 97 | ||
| JO620877.2 | - | 4 - 5 | 824 | 97 | ||
| JT716553.1 | - | 4 - 5 | 771 | 97 | ||
| JT738534.1 | - | 4 - 5 | 757 | 97 | ||
| KA333379.1 | - | 1 - 2 | 313 | 97 | ||
More Information
Set Membership: |
|

HSPA13
Genomic Map