Assay Details
| Gene Symbol: OSBPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675048.3 | XP_005675105.1 | 4 - 5 | 1261 | 64 | |
| GenBank mRNA | JO354114.2 | - | 4 - 5 | 410 | 64 | |
| JO391407.2 | - | 4 - 5 | 726 | 64 | ||
| JO428621.2 | - | 9 - 10 | 2094 | 64 | ||
| JO533510.2 | - | 4 - 5 | 410 | 64 | ||
| JO589364.2 | - | 4 - 5 | 390 | 64 | ||
| JO589365.2 | - | 4 - 5 | 754 | 64 | ||
| JT717569.1 | - | 10 - 11 | 1942 | 64 | ||
| JT737753.1 | - | 11 - 12 | 2254 | 64 | ||
| KA333604.1 | - | 9 - 10 | 2085 | 64 | ||
More Information
Set Membership: |
|

OSBPL11
Genomic Map