Assay Details
| Gene Symbol: C5H22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681063.3 | XP_005681120.1 | 5 - 6 | 656 | 79 | |
| XM_018048744.1 | XP_017904233.1 | 4 - 5 | 575 | 79 | ||
More Information
Set Membership: |
|

C5H22orf23
Genomic Map