Assay Details
| Gene Symbol: HOPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681640.3 | XP_005681697.1 | 2 - 3 | 728 | 89 | |
| XM_005681641.3 | XP_005681698.1 | 2 - 3 | 524 | 89 | ||
| XM_018049490.1 | XP_017904979.1 | 2 - 3 | 663 | 89 | ||
| XM_018049491.1 | XP_017904980.1 | 2 - 3 | 660 | 89 | ||
More Information
Set Membership: |
|

HOPX
Genomic Map