Assay Details
| Gene Symbol: MAPK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681861.3 | XP_005681918.1 | 11 - 12 | 1808 | 66 | |
| XM_005681862.2 | XP_005681919.1 | 11 - 12 | 1835 | 66 | ||
| XM_005681863.3 | XP_005681920.1 | 11 - 12 | 1808 | 66 | ||
| XM_013964768.2 | XP_013820222.1 | 12 - 13 | 1925 | 66 | ||
| XM_013964769.2 | XP_013820223.1 | 12 - 13 | 1742 | 66 | ||
| XM_013964770.2 | XP_013820224.1 | 11 - 12 | 1458 | 66 | ||
| XM_013964774.2 | XP_013820228.1 | 10 - 11 | 1736 | 66 | ||
| XM_018049689.1 | XP_017905178.1 | 11 - 12 | 1808 | 66 | ||
| XM_018049691.1 | XP_017905180.1 | 11 - 12 | 1808 | 66 | ||
| GenBank mRNA | JO634949.2 | - | 10 - 11 | 1713 | 66 | |
| JO634950.2 | - | 10 - 11 | 1712 | 66 | ||
More Information
Set Membership: |
|

MAPK10
Genomic Map