Assay Details
| Gene Symbol: LOC102182115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049575.1 | XP_017905064.1 | 3 - 4 | 410 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102182115| Gene Symbol: LOC102182115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049575.1 | XP_017905064.1 | 3 - 4 | 410 | 84 | |
Set Membership: |
|