Assay Details
| Gene Symbol: TPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683842.3 | XP_005683899.1 | 4 - 5 | 729 | 58 | |
| XM_018052137.1 | XP_017907626.1 | 4 - 5 | 729 | 58 | ||
| XM_018052138.1 | XP_017907627.1 | 4 - 5 | 729 | 58 | ||
| XM_018052139.1 | XP_017907628.1 | 4 - 5 | 729 | 58 | ||
| GenBank mRNA | JO538982.2 | - | 4 - 5 | 627 | 58 | |
More Information
Set Membership: |
|

TPM2
Genomic Map