Assay Details
| Gene Symbol: AHSA2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055395.1 | XP_017910884.1 | 5 - 6 | 693 | 99 | |
| XM_018055397.1 | XP_017910886.1 | 5 - 6 | 693 | 99 | ||
| XM_018055398.1 | XP_017910887.1 | 5 - 6 | 463 | 99 | ||
| XM_018055399.1 | XP_017910888.1 | 5 - 6 | 471 | 99 | ||
| XM_018055400.1 | XP_017910889.1 | 2 - 3 | 153 | 99 | ||
| XM_018055401.1 | XP_017910890.1 | 3 - 4 | 195 | 99 | ||
| XM_018055402.1 | XP_017910891.1 | 3 - 4 | 195 | 99 | ||
More Information
Set Membership: |
|

AHSA2P
Genomic Map