Assay Details
| Gene Symbol: MED22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968059.2 | XP_013823513.2 | 1 - 2 | 255 | 75 | |
| XM_018056141.1 | XP_017911630.1 | 3 - 4 | 362 | 75 | ||
| XM_018056142.1 | XP_017911631.1 | 3 - 4 | 331 | 75 | ||
| GenBank mRNA | JO429975.2 | - | 2 - 3 | 1538 | 75 | |
| JO577255.1 | - | 2 - 3 | 1608 | 75 | ||
More Information
Set Membership: |
|

MED22
Genomic Map