Assay Details
| Gene Symbol: PSMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692373.3 | XP_005692430.3 | 2 - 3 | 493 | 56 | |
| GenBank mRNA | JO580155.1 | - | 2 - 3 | 361 | 56 | |
| JO633545.2 | - | 5 - 6 | 1032 | 56 | ||
| KA309482.1 | - | 2 - 3 | 349 | 56 | ||
More Information
Set Membership: |
|

PSMD8
Genomic Map