Assay Details
| Gene Symbol: ALAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695886.3 | XP_005695943.1 | 10 - 11 | 1950 | 58 | |
| GenBank mRNA | JO421722.2 | - | 1 - 2 | 308 | 58 | |
| JO421723.2 | - | 1 - 2 | 308 | 58 | ||
More Information
Set Membership: |
|

ALAS1
Genomic Map