Assay Details
| Gene Symbol: PPP1R11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696636.3 | XP_005696693.1 | 1 - 2 | 287 | 72 | |
| GenBank mRNA | JO622842.2 | - | 1 - 2 | 338 | 72 | |
| Gene Symbol: RNF39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | KA305866.1 | - | 5 - 6 | 3415 | 72 | |
More Information
Set Membership: |
|

PPP1R11
Genomic Map