Assay Details
| Gene Symbol: PAK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696859.3 | XP_005696916.2 | 3 - 4 | 480 | 70 | |
| XM_005696860.3 | XP_005696917.2 | 3 - 4 | 474 | 70 | ||
| GenBank mRNA | KA329205.1 | - | 7 - 8 | 859 | 70 | |
More Information
Set Membership: |
|

PAK1IP1
Genomic Map