Assay Details
| Gene Symbol: ENPP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698709.3 | XP_005698766.2 | 6 - 7 | 1050 | 68 | |
| XM_013975396.2 | XP_013830850.2 | 6 - 7 | 1051 | 68 | ||
| XM_013975399.2 | XP_013830853.2 | 6 - 7 | 1051 | 68 | ||
| XM_018042101.1 | XP_017897590.1 | 6 - 7 | 1050 | 68 | ||
More Information
Set Membership: |
|

ENPP6
Genomic Map