Assay Details
| Gene Symbol: IL13RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044222.1 | XP_017899711.1 | 3 - 4 | 457 | 72 | |
| GenBank mRNA | JO425430.1 | - | 3278 | 72 | ||
More Information
Set Membership: |
|

IL13RA1
Genomic Map