Assay Details
| Gene Symbol: PRRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO418584.2 | - | 1 - 2 | 680 | 83 | |
| JO617602.2 | - | 1 - 2 | 304 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PRRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO418584.2 | - | 1 - 2 | 680 | 83 | |
| JO617602.2 | - | 1 - 2 | 304 | 83 | ||
Set Membership: |
|