Assay Details
| Gene Symbol: ZRSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005701066.3 | XP_005701123.3 | 1 - 2 | 77 | 57 | |
| GenBank mRNA | JT705418.1 | - | 11 - 12 | 1661 | 57 | |
More Information
Set Membership: |
|

ZRSR2
Genomic Map