Assay Details
| Gene Symbol: ZRSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005701066.3 | XP_005701123.3 | 9 - 10 | 874 | 90 | |
| GenBank mRNA | JT705418.1 | - | 3 - 4 | 739 | 90 | |
More Information
Set Membership: |
|

ZRSR2
Genomic Map