Assay Details
| Gene Symbol: MPIG6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709546.3 | XP_005709603.2 | 4 - 5 | 660 | 70 | |
| XM_018038624.1 | XP_017894113.1 | 4 - 5 | 678 | 70 | ||
| GenBank mRNA | JO340228.2 | - | 3 - 4 | 485 | 70 | |
| JO340229.2 | - | 3 - 4 | 485 | 70 | ||
More Information
Set Membership: |
|

MPIG6B
Genomic Map