Assay Details
| Gene Symbol: CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963947.2 | XP_013819401.1 | 7 - 8 | 971 | 65 | |
| GenBank mRNA | JO377083.2 | - | 7 - 8 | 808 | 65 | |
| JO646301.2 | - | 7 - 8 | 988 | 65 | ||
More Information
Set Membership: |
|

CS
Genomic Map