Assay Details
| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050416.1 | XP_017905905.1 | 3 - 4 | 483 | 58 | |
| XM_018050417.1 | XP_017905906.1 | 3 - 4 | 408 | 58 | ||
| GenBank mRNA | JO358216.2 | - | 3 - 4 | 472 | 58 | |
| JO581276.1 | - | 3 - 4 | 399 | 58 | ||
More Information
Set Membership: |
|

MBD3
Genomic Map