Assay Details
| Gene Symbol: LOC102176695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054881.1 | XP_017910370.1 | 12 - 13 | 1899 | 77 | |
| XM_018054882.1 | XP_017910371.1 | 13 - 14 | 1965 | 77 | ||
| GenBank mRNA | JO339437.2 | - | 12 - 13 | 2965 | 77 | |
| JO580143.1 | - | 10 - 11 | 2547 | 77 | ||
More Information
Set Membership: |
|

LOC102176695
Genomic Map