Assay Details
| Gene Symbol: LOC102176695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054881.1 | XP_017910370.1 | 3 - 4 | 736 | 76 | |
| XM_018054882.1 | XP_017910371.1 | 4 - 5 | 802 | 76 | ||
| GenBank mRNA | JO339437.2 | - | 4 - 5 | 807 | 76 | |
| JO580143.1 | - | 2 - 3 | 389 | 76 | ||
More Information
Set Membership: |
|

LOC102176695
Genomic Map