Assay Details
| Gene Symbol: TBC1D20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057663.1 | XP_017913152.1 | 3 - 4 | 510 | 61 | |
| GenBank mRNA | JO617809.2 | - | 3 - 4 | 480 | 61 | |
More Information
Set Membership: |
|

TBC1D20
Genomic Map