Assay Details
| Gene Symbol: DNAJC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969158.2 | XP_013824612.1 | 6 - 7 | 841 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJC5B| Gene Symbol: DNAJC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969158.2 | XP_013824612.1 | 6 - 7 | 841 | 68 | |
Set Membership: |
|