Assay Details
| Gene Symbol: RASSF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060091.1 | XP_017915580.1 | 4 - 5 | 1026 | 62 | |
| XM_018060092.1 | XP_017915581.1 | 3 - 4 | 860 | 62 | ||
More Information
Set Membership: |
|

RASSF5
Genomic Map